Methyl methylcarbamate 95%

Supplier: Ambeed
Danger

A132655-25G A132655-5G
77010-010EA 91.24 USD
77010-010 77010-012
Methyl methylcarbamate 95%
Methyl methylcarbamate

Precaución: for Research Use Only

Formula: C₃H₇NO₂
MW: 89.09 g/mol
MDL Number: MFCD00082210
CAS Number: 6642-30-4
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR