Nε-carbobenzoxy-L-lysine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Nε-Cbz-L-lysine, (S)-2-Amino-6-(((benzyloxy)carbonyl)amino)hexanoic acid, H-Lys(Z)-OH
A132643-1000G
A132643-100G
A132643-10G
A132643-25G
A132643-500G
A132643-5G
76996-468EA
451.95
USD
76996-468
76996-470
76996-472
76996-474
76996-476
76996-478
Nε-carbobenzoxy-L-lysine 98%
Nepsilon-carbobenzoxy-L-lysine
Precaución: for Research Use Only
Formula:
C₁₄H₂₀N₂O₄ MW: 280.32 g/mol |
MDL Number:
MFCD00002638 CAS Number: 1155-64-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...