(2-(Aminomethyl)phenyl)boronic acid hydrochloride 95%

Supplier: Ambeed
Warning

A132612-100MG A132612-1G A132612-250MG A132612-5G
76983-794EA 22 USD
76983-794 76983-796 76983-798 76983-800
(2-(Aminomethyl)phenyl)boronic acid hydrochloride 95%
(2-(Aminomethyl)phenyl)boronic acid hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₁BClNO₂
MDL Number: MFCD11504837
CAS Number: 850589-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR