(2-Methoxy-4-(methoxycarbonyl)phenyl)boronic acid 96%

Supplier: Ambeed
Warning

Synonyms: 2-Methoxy-4-(methoxycarbonyl)phenylboronic acid

A132438-1G A132438-250MG A132438-5G
77010-004EA 120 USD
77010-004 77010-006 77010-008
(2-Methoxy-4-(methoxycarbonyl)phenyl)boronic acid 96%
(2-Methoxy-4-(methoxycarbonyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BO₅
MW: 209.99 g/mol
MDL Number: MFCD13190192
CAS Number: 849758-14-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR