1-(Benzo[d]oxazol-2-yl)ethanone 97%

Supplier: Ambeed
Warning

A132383-100MG A132383-1G A132383-250MG
77027-108EA 16.98 USD
77027-108 77027-110 77027-112
1-(Benzo[d]oxazol-2-yl)ethanone 97%
1-(Benzo[d]oxazol-2-yl)ethanone

Precaución: for Research Use Only

Formula: C₉H₇NO₂
MW: 161.1574 g/mol
MDL Number: MFCD00223823
CAS Number: 122433-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR