1-Benzyl-3-bromopyrrolidin-2-one 97%

Supplier: Ambeed
Warning

A132369-100MG A132369-1G A132369-250MG
76983-784EA 66 USD
76983-784 76983-786 76983-788
1-Benzyl-3-bromopyrrolidin-2-one 97%
1-Benzyl-3-bromopyrrolidin-2-one

Precaución: for Research Use Only

Formula: C₁₁H₁₂BrNO
MDL Number: MFCD18089244
CAS Number: 77868-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR