Brusatol 98%

Supplier: Ambeed
Warning

Synonyms: (1R,2S,3S,3aS,3a1R,4R,6aR,7aR,11aS,11bR)-Methyl 1,2,9-trihydroxy-8,11a-dimethyl-4-((3-methylbut-2-enoyl)oxy)-5,10-dioxo-2,3,3a,4,5,6a,7,7a,10,11,11a,11b-dodecahydro-1H-3,3a1-(epoxymethano)dibenzo[de,g]chromene-3-carboxylate, (+)-Brusatol

A132366-1MG A132366-5MG
76983-780EA 37.14 USD
76983-780 76983-782
Brusatol 98%
Brusatol

Precaución: for Research Use Only

Formula: C₂₆H₃₂O₁₁
MW: 520.53 g/mol
MDL Number: MFCD01746220
CAS Number: 14907-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR