5-Bromo-4-chlorothiophene-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A132268-100MG A132268-250MG
76983-776EA 75.33 USD
76983-776 76983-778
5-Bromo-4-chlorothiophene-2-carboxylic acid 97%
5-Bromo-4-chlorothiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₂BrClO₂S
MW: 241.49 g/mol
MDL Number: MFCD08694669
CAS Number: 123418-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR