(R)-2-Amino-2-(3-chlorophenyl)ethanol 97%

Supplier: Ambeed
Warning

A132233-100MG A132233-1G A132233-250MG
76996-444EA 37.14 USD
76996-444 76996-446 76996-448
(R)-2-Amino-2-(3-chlorophenyl)ethanol 97%
(R)-2-Amino-2-(3-chlorophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₁₀ClNO
MW: 171.62 g/mol
MDL Number: MFCD06659478
CAS Number: 926291-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR