4-Amino-6-chlorobenzene-1,3-disulfonamide 97%

Supplier: Ambeed
Warning

A132024-100G A132024-10G A132024-1G A132024-25G A132024-500G A132024-5G
76996-430EA 31 USD
76996-430 76996-432 76996-434 76996-436 76996-438 76996-440
4-Amino-6-chlorobenzene-1,3-disulfonamide 97%
4-Amino-6-chlorobenzene-1,3-disulphonamide

Precaución: for Research Use Only

Formula: C₆H₈ClN₃O₄S₂
MDL Number: MFCD00007933
CAS Number: 121-30-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR