tert-Butyl 4-(1H-indazol-3-yl)piperidine-1-carboxylate 95%
Supplier: Ambeed
|
Warning
|
A131943-100MG
A131943-1G
A131943-250MG
76983-764EA
51.99
USD
76983-764
76983-766
76983-768
tert-Butyl 4-(1H-indazol-3-yl)piperidine-1-carboxylate 95%
tert-Butyl 4-(1H-indazol-3-yl)piperidine-1-carboxylate
Precaución: for Research Use Only
Formula:
C₁₇H₂₃N₃O₂ MW: 301.38 g/mol |
CAS Number:
889945-69-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...