2-Chlorobenzo[d]thiazole-7-carboxylic acid 95%

Supplier: Ambeed
Warning

A131908-100MG A131908-1G A131908-250MG A131908-5G
76983-756EA 107.16 USD
76983-756 76983-758 76983-760 76983-762
2-Chlorobenzo[d]thiazole-7-carboxylic acid 95%
2-Chlorobenzo[d]thiazole-7-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₄ClNO₂S
MW: 213.64 g/mol
MDL Number: MFCD13195420
CAS Number: 1379324-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR