(1-Ethyl-1H-indazol-5-yl)boronic acid 95%

Supplier: Ambeed
Warning

A131891-100MG A131891-250MG
76983-752EA 48.81 USD
76983-752 76983-754
(1-Ethyl-1H-indazol-5-yl)boronic acid 95%
(1-Ethyl-1H-indazol-5-yl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BN₂O₂
MW: 190.01 g/mol
MDL Number: MFCD22576144
CAS Number: 952319-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR