3-Aminopiperidin-2-one 98%

Supplier: Ambeed
Warning

A131861-1G A131861-5G A131861-250MG A131861-25G
77027-090EA 38 USD
77027-090 77599-898 77666-898 77871-613
3-Aminopiperidin-2-one 98%
3-Aminopiperidin-2-one

Precaución: for Research Use Only

Formula: C5H10N2O
MDL Number: MFCD06809610
CAS Number: 1892-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR