(S)-Methyl 4-(1-aminoethyl)benzoate hydrochloride 97%

Supplier: Ambeed
Warning

A131676-1G A131676-250MG A131676-5G
76983-740EA 9.55 USD
76983-740 76983-742 76983-744 76983-746
(S)-Methyl 4-(1-aminoethyl)benzoate hydrochloride 97%
(S)-Methyl 4-(1-aminoethyl)benzoate hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₄ClNO₂
MDL Number: MFCD11505983
CAS Number: 847728-91-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR