Olmesartan medoxomil 98%

Supplier: Ambeed
Warning

Synonyms: (5-Methyl-2-oxo-1,3-dioxol-4-yl)methyl 1-((2'-(2H-tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)-4-(2-hydroxypropan-2-yl)-2-propyl-1H-imidazole-5-carboxylate, (5-Methyl-2-oxo-1,3-dioxol-4-yl)methyl-1-((2'-(1H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl)methyl)-4-(2-hydroxypropan-2-yl)-2-propyl-1H-imidazole-5-carboxylate

A131632-100MG A131632-10MG A131632-1MG A131632-250MG A131632-50MG A131632-5MG A131632-100G A131632-25G
76996-406EA 23.34 USD
76996-406 76996-408 76996-410 76996-412 76996-414 76996-416 77670-860 77670-862
Olmesartan medoxomil 98%
Olmesartan medoxomil

Precaución: for Research Use Only

Formula: C₂₉H₃₀N₆O₆
MW: 558.60 g/mol
MDL Number: MFCD00944911
CAS Number: 144689-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR