(R)-tert-Butyl 3-aminopyrrolidine-1-carboxylate hydrochloride 95%

Supplier: Ambeed
Warning

A131627-10G A131627-1G A131627-25G
77027-084EA 40.32 USD
77027-084 77027-086 77027-088
(R)-tert-Butyl 3-aminopyrrolidine-1-carboxylate hydrochloride 95%
(R)-tert-Butyl 3-aminopyrrolidine-1-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₉ClN₂O₂
MW: 222.71 g/mol
MDL Number: MFCD08061978
CAS Number: 1004538-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR