4-(Bromomethyl)-2-fluorobenzonitrile 98%, Ambeed.Inc

Supplier: Ambeed
Danger

Synonyms: 4-Cyano-3-fluorobenzyl bromide, α-Bromo-2-fluoro-p-tolunitrile

A131577-100MG A131577-10G A131577-1G A131577-250MG A131577-25G A131577-5G
77009-978EA 6 USD
77009-978 77009-980 77009-982 77009-984 77009-986 77683-167
4-(Bromomethyl)-2-fluorobenzonitrile 98%, Ambeed.Inc
4-(Bromomethyl)-2-fluorobenzonitrile

Precaución: For research use only.

Formula: C₈H₅BrFN
MDL Number: MFCD03094323
CAS Number: 222978-03-2
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR