D-Valine methyl ester hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A131523-100G
A131523-10G
A131523-1G
A131523-25G
A131523-500G
A131523-5G
76996-390EA
50
USD
76996-390
76996-392
76996-394
76996-396
76996-398
77599-894
D-Valine methyl ester hydrochloride 95%
D-Valine methyl ester hydrochloride
Precaución: for Research Use Only
Formula:
C₆H₁₃NO₂ |
MDL Number:
MFCD00237309 CAS Number: 7146-15-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...