2-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol 98%

Supplier: Ambeed
Warning

A131309-100MG A131309-1G A131309-250MG A131309-5G
76983-712EA 15 USD
76983-712 76983-714 76983-716 76983-718
2-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol 98%
2-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

Precaución: for Research Use Only

Formula: C₁₃H₁₉BO₃
MDL Number: MFCD16994279
CAS Number: 627906-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR