5-Nitro-2-thiophenecarboxylic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Nitrothiophene-2-carboxylic acid
A131259-100G
A131259-25G
A131259-5G
A131259-1G
76983-706EA
179.3
USD
76983-706
76983-708
76983-710
77683-165
5-Nitro-2-thiophenecarboxylic acid 95%
5-Nitro-2-thiophenecarboxylic acid
Precaución: For research use only.
Formula:
C₅H₃NO₄S MW: 173.14 g/mol |
MDL Number:
MFCD00159552 CAS Number: 6317-37-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...