4-Amino-m-cresol 97%

Supplier: Ambeed
Warning

A131170-100G A131170-10G A131170-25G
76996-382EA 31 USD
76996-382 76996-384 77298-456
4-Amino-m-cresol 97%
4-Amino-m-cresol

Precaución: for Research Use Only

Formula: C₇H₉NO
MDL Number: MFCD00007871
CAS Number: 2835-99-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR