1-(4-Bromophenyl)cyclopropanecarboxylic acid 98%

Supplier: Ambeed
Warning

A131163-1G A131163-250MG A131163-5G A131163-10G
76983-694EA 6.37 USD
76983-694 76983-696 76983-698 77611-162
1-(4-Bromophenyl)cyclopropanecarboxylic acid 98%
1-(4-Bromophenyl)cyclopropanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₉BrO₂
MW: 241.0800 g/mol
MDL Number: MFCD07374439
CAS Number: 345965-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR