2-(3-(4-Chlorophenyl)pyrrolidin-3-yl)ethanol 95%

Supplier: Ambeed
Warning

A131153-100MG A131153-1G A131153-250MG
77009-952EA 39 USD
77009-952 77009-954 77009-956
2-(3-(4-Chlorophenyl)pyrrolidin-3-yl)ethanol 95%
2-(3-(4-Chlorophenyl)pyrrolidin-3-yl)ethanol

Precaución: for Research Use Only

Formula: C₁₂H₁₆ClNO
MDL Number: MFCD17014882
CAS Number: 52423-70-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR