Ethyl-5-nitro-1H-pyrazole-3-carboxylate 98%

Supplier: Ambeed
Warning

A131082-100MG A131082-1G A131082-250MG A131082-25G A131082-5G
77027-060EA 8 USD
77027-060 77027-062 77027-064 77027-066 77027-068
Ethyl-5-nitro-1H-pyrazole-3-carboxylate 98%
Ethyl-5-nitro-1H-pyrazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₆H₇N₃O₄
MDL Number: MFCD04971172
CAS Number: 39846-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR