tert-Butyl 4-(2-cyano-4-nitrophenyl)piperazine-1-carboxylate 97%

Supplier: Ambeed
Warning

A130989-1G A130989-250MG A130989-5G
76983-688EA 99.73 USD
76983-688 76983-690 76983-692
tert-Butyl 4-(2-cyano-4-nitrophenyl)piperazine-1-carboxylate 97%
tert-Butyl 4-(2-cyano-4-nitrophenyl)piperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₆H₂₀N₄O₄
MW: 332.35 g/mol
MDL Number: MFCD26383922
CAS Number: 288251-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR