4-Benzyl-1-oxa-4,9-diazaspiro[5.5]undecan-3-one 97%

Supplier: Ambeed
Warning

A130902-100MG A130902-250MG
76983-684EA 128 USD
76983-684 76983-686
4-Benzyl-1-oxa-4,9-diazaspiro[5.5]undecan-3-one 97%
4-Benzyl-1-oxa-4,9-diazaspiro[5.5]undecan-3-one

Precaución: for Research Use Only

Formula: C₁₅H₂₀N₂O₂
MDL Number: MFCD16619254
CAS Number: 151096-96-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR