(2-Chloro-4-nitrophenoxy)acetic acid 95+%

Supplier: Ambeed
Warning

A130900-1G A130900-5G
77027-056EA 11 USD
77027-056 77027-058
(2-Chloro-4-nitrophenoxy)acetic acid 95+%
(2-Chloro-4-nitrophenoxy)acetic acid

Precaución: for Research Use Only

Formula: C₈H₆ClNO₅
MDL Number: MFCD03414519
CAS Number: 5037-04-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR