Ethyl 2-bromo-5-chloro-1,3-thiazole-4-carboxylate 98%

Supplier: Ambeed
Warning

A130891-1G A130891-250MG
77027-052EA 118.83 USD
77027-052 77027-054
Ethyl 2-bromo-5-chloro-1,3-thiazole-4-carboxylate 98%
Ethyl 2-bromo-5-chloro-1,3-thiazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₆H₅BrClNO₂S
MDL Number: MFCD08704625
CAS Number: 425392-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR