Ethyl-1H-indazole-5-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl Indazole-5-carboxylate, Ethyl 1H-indazole-5-carboxylate, Indazole-5-carboxylic acid ethyl ester

A130885-1G A130885-250MG A130885-5G
77009-924EA 29.71 USD
77009-924 77009-926 77009-928
Ethyl-1H-indazole-5-carboxylate 95%
Ethyl 1H-indazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₂
MW: 190.20 g/mol
MDL Number: MFCD03426220
CAS Number: 192944-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR