Methyl (2S)-morpholine-2-carboxylate hydrochloride 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-Methyl morpholine-2-carboxylate hydrochloride
A130819-100MG
A130819-10G
A130819-250MG
A130819-5G
A130819-1G
77009-916EA
39.26
USD
77009-916
77009-918
77009-920
77009-922
77611-158
Methyl (2S)-morpholine-2-carboxylate hydrochloride 95%
Methyl (2S)-morpholine-2-carboxylate hydrochloride
Precaución: for Research Use Only
Formula:
C₆H₁₁NO₃·ClH MW: 181.617 g/mol |
MDL Number:
MFCD21648490 CAS Number: 1417789-45-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...