(±)-2-Aminobutyric acid 95%

Supplier: Ambeed
Warning

A130614-10G A130614-25G A130614-500G A130614-5G A130614-100G
76996-364EA 6.37 USD
76996-364 76996-366 76996-368 76996-370 77670-846
(±)-2-Aminobutyric acid 95%
(±)-2-Aminobutyric acid

Precaución: for Research Use Only

Formula: C₄H₉NO₂
MW: 103.12 g/mol
MDL Number: MFCD00008093
CAS Number: 2835-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR