(1-Hydroxy-2-(imidazo[1,2-a]pyridin-3-yl)ethane-1,1-diyl)diphosphonic acid 95%

Supplier: Ambeed
Warning

Synonyms: Minodronic acid

A130596-100MG A130596-1G A130596-5G
76983-674EA 59.42 USD
76983-674 76983-676 76983-678
(1-Hydroxy-2-(imidazo[1,2-a]pyridin-3-yl)ethane-1,1-diyl)diphosphonic acid 95%
(1-Hydroxy-2-(imidazo[1,2-a]pyridin-3-yl)ethane-1,1-diyl)diphosphonic acid

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O₇P₂
MW: 322.14 g/mol
MDL Number: MFCD00890772
CAS Number: 180064-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR