[1,2,4]Triazolo[1,5-a]pyridine-6-carbonitrile 95%

Supplier: Ambeed
Warning

A130573-100MG A130573-1G A130573-250MG A130573-5G
77009-902EA 6 USD
77009-902 77009-904 77009-906 77009-908
[1,2,4]Triazolo[1,5-a]pyridine-6-carbonitrile 95%
[1,2,4]Triazolo[1,5-a]pyridine-6-carbonitrile

Precaución: for Research Use Only

Formula: C₇H₄N₄
MDL Number: MFCD22093011
CAS Number: 943845-23-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR