tert-Butyl 3-((dimethylamino)methylene)-4-oxopyrrolidine-1-carboxylate 97%

Supplier: Ambeed
Warning

A130506-1G A130506-250MG A130506-5G
76983-666EA 44.56 USD
76983-666 76983-668 76983-670
tert-Butyl 3-((dimethylamino)methylene)-4-oxopyrrolidine-1-carboxylate 97%
tert-Butyl 3-((dimethylamino)methylene)-4-oxopyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₂₀N₂O₃
MW: 240.29 g/mol
MDL Number: MFCD11616027
CAS Number: 157327-42-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR