4-(Boc-amino)-3-iodopyridine 98%

Supplier: Ambeed
Warning

A130495-10G A130495-1G A130495-250MG A130495-5G
77009-888EA 51 USD
77009-888 77009-890 77009-892 77009-894
4-(Boc-amino)-3-iodopyridine 98%
4-(Boc-amino)-3-iodopyridine

Precaución: for Research Use Only

Formula: C₁₀H₁₃IN₂O₂
MDL Number: MFCD04973409
CAS Number: 211029-67-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR