Fmoc-L-allylglycine 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Gly(allyl)-OH

A130423-100G A130423-10G A130423-1G A130423-25G A130423-5G
76996-352EA 975 USD
76996-352 76996-354 76996-356 76996-358 76996-360 76996-362
Fmoc-L-allylglycine 97%
Fmoc-L-allylglycine

Precaución: for Research Use Only

Formula: C₂₀H₁₉NO₄
MW: 337.38 g/mol
MDL Number: MFCD01311749
CAS Number: 146549-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR