3-Amino-3-oxopropanoic acid 95%

Supplier: Ambeed
Warning

A130194-100MG A130194-250MG A130194-5G A130194-1G A130194-25G
77027-028EA 37 USD
77027-028 77027-030 77027-032 77670-842 77871-607
3-Amino-3-oxopropanoic acid 95%
3-Amino-3-oxopropanoic acid

Precaución: for Research Use Only

Formula: C₃H₅NO₃
MDL Number: MFCD00154605
CAS Number: 2345-56-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR