2-Iodoresorcinol 98%

Supplier: Ambeed
Warning

Synonyms: 1,3-Dihydroxy-2-iodobenzene, 2-Iodobenzene-1,3-diol

A130161-100MG A130161-1G A130161-250MG A130161-5G
77027-014EA 41.38 USD
77027-014 77027-016 77027-018 77027-020
2-Iodoresorcinol 98%
2-Iodoresorcinol

Precaución: for Research Use Only

Formula: C₆H₅IO₂
MW: 236.01 g/mol
MDL Number: MFCD09701440
CAS Number: 41046-67-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR