2-Bromo-N,N,4-trimethylaniline 97%

Supplier: Ambeed
Warning

A130093-10G A130093-1G A130093-5G
76983-654EA 480.59 USD
76983-654 76983-656 76983-658
2-Bromo-N,N,4-trimethylaniline 97%
2-Bromo-N,N,4-trimethylaniline

Precaución: for Research Use Only

Formula: C₉H₁₂BrN
MW: 214.10 g/mol
MDL Number: MFCD07780654
CAS Number: 23667-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR