3-Bromo-2-fluorophenylacetic acid 98%
Supplier: Ambeed
|
Warning
|
A129988-100MG
A129988-1G
A129988-250MG
A129988-5G
A129988-25G
76983-646EA
19.1
USD
76983-646
76983-648
76983-650
77683-154
77683-153
3-Bromo-2-fluorophenylacetic acid 98%
3-Bromo-2-fluorophenylacetic acid
Precaución: For research use only.
Formula:
C8H6BrFO2 MW: 233.0300 g/mol |
MDL Number:
MFCD09864663 CAS Number: 1375964-39-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...