3-Methyl-5-nitro-1H-pyrazole 97%

Supplier: Ambeed
Warning

A129730-100G A129730-10G A129730-1G A129730-25G A129730-5G
76996-292EA 269.47 USD
76996-292 76996-294 76996-296 76996-298 76996-300
3-Methyl-5-nitro-1H-pyrazole 97%
3-Methyl-5-nitro-1H-pyrazole

Precaución: for Research Use Only

Formula: C4H5N3O2
MW: 127.1014 g/mol
MDL Number: MFCD00463977
CAS Number: 34334-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR