4-(Pyrimidin-2-yl)morpholine ≥95%

Supplier: Ambeed
Warning

A129651-100MG A129651-250MG
77009-856EA 47.75 USD
77009-856 77009-858
4-(Pyrimidin-2-yl)morpholine ≥95%
4-(Pyrimidin-2-yl)morpholine

Precaución: for Research Use Only

Formula: C₈H₁₁N₃O
MW: 165.19 g/mol
MDL Number: MFCD27925725
CAS Number: 57356-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR