3-Amino-5-bromo-1-methylpyridin-2(1H)-one 95%

Supplier: Ambeed
Warning

A129636-10G A129636-1G A129636-25G A129636-5G
76983-592EA 93 USD
76983-592 76983-594 76983-596 76983-598
3-Amino-5-bromo-1-methylpyridin-2(1H)-one 95%

Precaución: for Research Use Only

Formula: C₆H₇BrN₂O
MDL Number: MFCD09835082
CAS Number: 910543-72-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR