Methyl-1H-indazole-6-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 1H-indazole-6-carboxylate, Indazole-6-carboxylic acid methyll ester, Methyl Indazole-6-carboxylate

A129542-100G A129542-10G A129542-1G A129542-25G A129542-500G A129542-5G
77009-836EA 267.35 USD
77009-836 77009-838 77009-840 77009-842 77009-844 77009-846
Methyl-1H-indazole-6-carboxylate 97%
Methyl 1H-indazole-6-carboxylate

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MW: 176.17 g/mol
MDL Number: MFCD07371612
CAS Number: 170487-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR