(R)-2-Amino-3-(tritylthio)propanamide 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-Cys(Trt)-NH2, H-Cys(Trt)-NH₂, S-Trityl-L-cysteinamide
A129329-1G
A129329-250MG
A129329-5G
76996-272EA
26
USD
76996-272
76996-274
76996-276
(R)-2-Amino-3-(tritylthio)propanamide 95%
(R)-2-Amino-3-(tritylthio)propanamide
Precaución: for Research Use Only
Formula:
C₂₂H₂₂N₂OS |
MDL Number:
MFCD22126061 CAS Number: 166737-85-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...