5-Bromoquinazoline 97%

Supplier: Ambeed
Warning

A129328-100MG A129328-1G A129328-250MG A129328-5G
77026-988EA 20 USD
77026-988 77026-990 77026-992 77026-994
5-Bromoquinazoline 97%
5-Bromoquinazoline

Precaución: for Research Use Only

Formula: C₈H₅BrN₂
MDL Number: MFCD11036107
CAS Number: 958452-00-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR