1-Cyclopropyl-6-fluoro-7-chloro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 7-Chloro-1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-1,8-naphthyridine-3-carboxylic acid

A129312-25G A129312-5G A129312-100G
77026-980EA 61 USD
77026-980 77026-982 77871-598
1-Cyclopropyl-6-fluoro-7-chloro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid 98%
1-Cyclopropyl-6-fluoro-7-chloro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₈ClFN₂O₃
MW: 282.6600 g/mol
MDL Number: MFCD08458865
CAS Number: 100361-18-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR