3-(4-Ethylphenyl)propionic acid 98%

Supplier: Ambeed
Warning

A129238-1G A129238-250MG A129238-5G
76996-266EA 28 USD
76996-266 76996-268 76996-270
3-(4-Ethylphenyl)propionic acid 98%
3-(4-Ethylphenyl)propionic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
MDL Number: MFCD03425687
CAS Number: 64740-36-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR