N-Carbobenzoxy-D-leucine 95%

Supplier: Ambeed
Warning

Synonyms: Z-D-Leu-OH, N-Cbz-D-leucine

A129217-100G A129217-10G A129217-1G A129217-25G A129217-5G
76996-256EA 196.27 USD
76996-256 76996-258 76996-260 76996-262 76996-264
N-Carbobenzoxy-D-leucine 95%
N-Carbobenzoxy-D-leucine

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₄
MW: 265.31 g/mol
MDL Number: MFCD00066068
CAS Number: 28862-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR